Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Manufacturer: OmnifitSupplier Diversity Partner 006SFE15
 View more versions of this product
Catalog No. 14-500-698
  Check Availability
Add to Cart
    Specifications
    For Use With Omnifit SolventPlus chromatography columns
    Material PTFE
    Diameter (Metric) Inner 15mm
    Diameter (Metric) Outer 15mm
    Type Replacement fixed endpieces for SolventPlus column
  • Description & Specifications
Related Products
Product Content Feedback