Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Manufacturer: OmnifitSupplier Diversity Partner 006SCC0610FF
 View more versions of this product
Catalog No. 14-500-647
  Check Availability
Add to Cart
    Specifications
    For Use With Omnifit SolventPlus chromatography columns
    Length (Metric) 100mm
    Diameter (Metric) Inner 6.6mm
    Diameter (Metric) Outer 6.6mm
    Includes Glass column assembly, frit kit, fittings kit; Two fixed endpieces
    Type SolventPlus chromatography column
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback