Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Manufacturer: OmnifitSupplier Diversity Partner 006SCC1015AF
 View more versions of this product
Catalog No. 14-500-654
  Check Availability
Add to Cart
    Specifications
    For Use With Omnifit SolventPlus chromatography columns
    Length (Metric) 150mm
    Diameter (Metric) Inner 10mm
    Diameter (Metric) Outer 10mm
    Includes Glass column assembly, frit kit, fittings kit; One adjustable, one fixed endpiece
    Type SolventPlus chromatography column
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback