Learn More
Kinesis™ Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Manufacturer:  Kinesis™ 006SCC0605AA
 View more versions of this product
Catalog No. 14-500-631
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Borosilicate glass and PTFE construction, with Chemraz™ O-rings and PTFE frits put only inert materials in the fluid path
  • ETFE end caps and fitting nuts ensure a chemically inert connection into and out of the system
  • PTFE endpiece makes the primary seal to glass and has even distribution pattern to give even sample application across the bed surface
  • Ready-assembled column includes fixed or adjustable endpieces (-FF, -AF or -AA configuration) and 30µm PE frits
Specifications
Specifications
Omnifit SolventPlus chromatography columns
50mm
6.6mm
SolventPlus chromatography column
6.6mm
Glass column assembly with PTFE endpiece, frit, 2 Adjustable Endpieces
Documents
Accessories