Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Omnipore™ PTFE Membrane Filter

$424.50

Specifications

Color White
Filter Type Screen filter
For Use With (Application) Compatible with virtually all solvents, acids and alkaline solutions
Membrane PTFE
Type Membrane Filter
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (Metric) Filter Pore Size Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Filter Pore Size Diameter (Metric) Outer Price Quantity & Availability  
JHWP14225
View Documents
MilliporeSigma™
JHWP14225
142mm 0.45μm 142 mm Each for $424.50
Add to cart
 
Specifications

Specifications

White
Compatible with virtually all solvents, acids and alkaline solutions
Membrane Filter
Screen filter
PTFE
PTFE
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit