Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ OmniPur™ Coomassie™ Blue R-250

Used for post-electrophoresis staining of proteins.

Supplier:  MilliporeSigma™ 334025GM

 View more versions of this product

Catalog No. 33-402-5GM

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

EMD Millipore OmniPur™ Coomassie Blue R-250 for applications requiring a high degree of sensitivity solubility.

  • Use for post-electrophoresis staining of proteins in applications requiring high degree of sensitivity or solubility
  • CAS: 6104-59-2
  • Molecular formula: C45H44N3NaO7S2
Specifications

Specifications

Coomassie Blue R-250 Powder
Blue
Protein Stain
25 g
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit