Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OmniPur™ Glycine, MilliporeSigma™
Click to view available options
Quantity:
5 kg
Specifications
CAS | 56-40-6 |
Molecular Formula | H2NCH2 |
Synonym | Aminoacetic Acid |
Packaging | Plastic Drum |
Physical Form | Crystalline powder |
Percent Purity | ≥ 98.5% by assay |
Color | White |
Quantity | 5 kg |
Safety and Handling
Recommended Storage : Store at room temperature