Learn More
OMP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00037W
Catalog No. PIOSR00037W
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
In IHC applications, Zamboni′s or 4% PFA fixatives are recommended. Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OMP
Polyclonal
OMP
Olfactory neuronal-specific protein
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
P47874
OMP
Synthetic peptide from human Olfactory Marker Protein.
RUO
4975
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations