Learn More
Azzota Corp ON-123300 DFS Item
Manufacturer:  Azzota Corp C1403
ON-123300 is a potent multikinase inhibitor of ARK5 and CDK4, as well as growth factor receptor tyrosine kinases such as β-type platelet-derived growth factor receptor (PDGFRβ). Please request if need more quantities
Catalog No. 50-161-3211