Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oncomodulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121725

This item is not returnable.
View return policy
Description
Oncomodulin Polyclonal antibody specifically detects Oncomodulin in Human, Mouse samples. It is validated for Western BlotSpecifications
Oncomodulin | |
Polyclonal | |
Unconjugated | |
Ocm | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
P0CE72, P51879 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
Ocm | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title