Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oncostatin M/OSM Mouse anti-Human, Mouse, Alexa Fluor™ 350, Clone: 17001.31, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB12010842AF350

Catalog No. N1200142A30


Add to cart

Description

Description

Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Oncostatin M/OSM
17001.31
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
OSM
Recombinant full length protein (Human) expressed in E. coli.
Protein A purified
Biologically Active Proteins, Cell Cycle and Replication
Primary
5008.0
Human, Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
MGC20461, oncostatin M, oncostatin-M
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Does not cross-react with recombinant human IL-6, IL-11, CNTF, LIF and recombinant mouse OSM.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit