Learn More
Apexbio Technology LLC Ondansetron HCl 99614-01-4 10mg
SDP
Supplier:  Apexbio Technology LLC A516610
Encompass_Preferred
Ondansetron HCl (CAS 99614-01-4) is a selective antagonist of the serotonin 5-hydroxytryptamine 3 (5-HT3) receptor By inhibiting the activation of these ligand-gated ion channels it modulates serotonergic signaling pathways implicated in emesis and other neuropharmacological processes Ondansetron HCl is widely utilized in biomedical research to investigate serotonergic modulation in gastrointestinal central nervous system and chemotherapy-induced emesis models Its specific inhibition of 5-HT3 receptors makes it a valuable tool for delineating the physiological and pharmacological roles of this receptor subtype across various experimental paradigms
Catalog No. 50-000-97335