Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ondansetron hydrochloride dihydrate, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
103639-04-9
HCl·2H2O
365.85
BGGIFKYQGVGSIW-UHFFFAOYSA-N
1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
71317260
chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]
103639-04-9
HCl·2H2O
365.85
BGGIFKYQGVGSIW-UHFFFAOYSA-N
1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
71317260
chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]
Melting Point 178.0°C
Infrared Spectrum Authentic
Water 10.5% max.
Quantity 5g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 365.85
Percent Purity 98%
Chemical Name or Material Ondansetron hydrochloride dihydrate

GHS H Statement Toxic if swallowed. Toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger