Learn More
Brady™ One-Sided Polyester Tags: INSPECTED
Identify facility equipment to your exact specifications. Brady™ One-Sided Polyester Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 76157
Catalog No. 19-113-135
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
25/Pack
-40°F to 212°F
INSPECTED
Black on white
General equipment
Rectangle
Polyester
OSHA 1910.145
3 in. x 5.75 in.
(25) nylon fasteners