Learn More
ONECUT3 Mouse anti-Human, Alexa Fluor 405, Clone: 10B2F5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237340AF405
Catalog No. NBP237340N
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ONECUT3
10B2F5
Western Blot, ELISA, Immunohistochemistry
Purified
ONECUT3
Purified recombinant fragment of human ONECUT3 (AA: 350-455) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
OC3, OC-3, one cut domain family member 3, one cut domain, family member 3, one cut homeobox 3, transcription factor ONECUT-3
Mouse
IgG1
0.1 ml
Primary
390874