Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ONECUT3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548948

Catalog No. PA548948

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Transcriptional activator. Binds the consensus DNA sequence 5 ft.-DHWATTGAYTWWD-3 ft. on a variety of gene promoters such as those of HNF3B and TTR.
Specifications

Specifications

ONECUT3
Polyclonal
PBS with 0.09% sodium azide
O60422
One cut domain family member 3, One cut homeobox 3, Transcription factor ONECUT-3, OC-3, ONECUT3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
390874
Western Blot
Unconjugated
ONECUT3
Liquid
ONECUT3
KLH conjugated synthetic peptide between 393-419 amino acids from the C-terminal region of human ONECUT3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit