Onguard™ Blue Max™ Work Boots

Moderate chemical resistance for work in poultry and food processing

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 891021400

 View more versions of this product

Catalog No. 19-806-635

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM
    For Use With (Application) Excellent for use in poultry, seafood processing, food processing and canneries
    Gender Male
    Height (English) 16 in.
    Shank Material Steel
    Sole Features Replaceable cushion insoles, slip resistance, Ultragrip™ sipe outsole
    Toe Style Steel
    Color Blue
    Material PVC
    Size 14

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit