Learn More
Dunlop™ Onguard™ Buckle Arctic Boots
Waterproof and durable rubber
Supplier: Dunlop™ 860630700
This item is not returnable.
View return policy
Description
- Provide maximum protection against the elements
- 11 in. high (28cm) boots fit over most work shoes and boots
- Feature cleated soles, four noncorrosive buckles, stretch-net lining inside and self-draining gusset
Specifications
Black | |
11 in. | |
Rubber | |
Stretch net lining, self cleaning, self draining gusset, four noncorrosive buckles, cleated soles |
Construction, utilities, roadwork and foul weather | |
Cotton | |
7 |