Onguard™ Buckle Arctic Boots

Waterproof and durable rubber

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 860630800

 View more versions of this product

Catalog No. 19-024395

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Construction, utilities, roadwork and foul weather
    Height (English) 11 in.
    Sole Features Stretch net lining, self cleaning, self draining gusset, four noncorrosive buckles, cleated soles
    Color Black
    Material Rubber
    Size 8
    Liner Material Cotton

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit