Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Onguard Budget Bib Overalls

Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 73552XL00

 View more versions of this product

Catalog No. 19-120-896D


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Snap fly
Yellow
130.8cm
PVC
X-Large
Budget
Lightweight applications
Adjustable snap
78.7cm
Take up snap
2-ply
51.5 in.
2.0
50 in.
Adjustable reinforced
0.2mm
31 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit