Learn More
Onguard™ Buffalo PVC Work Boots
Ideal for use in general industrial safety applications
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 87801 14
 View more versions of this product
Catalog No. 19-099-159M
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM F2413-11, ANSI Z41 PT99 M I/75 C/75
Used in construction, farming, agriculture and aquaculture
16 in.
V-bar molded
Steel
Yes
PVC
Pull-on
Male
Kick off
Self-cleaning lug outsole for traction in wet, muddy conditions
Safety steel
Black
14