Learn More
Dunlop™ Onguard™ Buffalo PVC Work Boots
Ideal for use in general industrial safety applications
Manufacturer:  Dunlop™ 87801 7
 View more versions of this product
Catalog No. 19-099-159E
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Economical and durable, these PVC injection-molded boots offer high abrasion resistance
  • Feature a lug outsole for sure traction and superior grip in wet, muddy conditions
  • V-bar molded shank and wide shaft enhance worker comfort
  • Molded-in insole
  • Net lined for easy on and off
Order Info
Available with steel toe or plain toe
Compliance
Boots meet or exceed ASTM™ F2413-11 specifications.
Specifications
Specifications
ASTM F2413-11, ANSI Z41 PT99 M I/75 C/75
Pull-on
16 in.
PVC
7
Safety Steel Toe
Black
Used in construction, farming, agriculture and aquaculture
Kick-off
V-bar molded
Steel