Learn More
Dunlop™ Onguard™ Buffalo PVC Work Boots
Ideal for use in general industrial safety applications
Supplier: Dunlop™ 87401 5
This item is not returnable.
View return policy
Description
- Economical and durable, these PVC injection-molded boots offer high abrasion resistance
- Feature a lug outsole for sure traction and superior grip in wet, muddy conditions
- V-bar molded shank and wide shaft enhance worker comfort
- Molded-in insole
- Net lined for easy on and off
Order Info
Available with steel toe or plain toe
Compliance
Boots meet or exceed ASTM™ F2413-11 specifications.
Specifications
ASTM F2413-11, ANSI Z41 PT99 M I/75 C/75 | |
Pull-on | |
16 in. | |
PVC | |
5 | |
Plain |
Black | |
Used in construction, farming, agriculture and aquaculture | |
Kick-off | |
V-bar molded | |
Self-cleaning lug outsole for traction in wet, muddy conditions | |
Yes |