Onguard™ Buffalo PVC Work Boots

Ideal for use in general industrial safety applications

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 87401 13

 View more versions of this product

Catalog No. 19-099-160L

  Check Availability
Add to Cart

    Specifications

    Gender Male
    Lug Type Kick off
    Toe Material Plain
    Waterproof Yes
    Material PVC
    Certifications/Compliance ASTM F2413-11, ANSI Z41 PT99 M I/75 C/75
    Closure Type Pull-on
    For Use With (Application) Used in construction, farming, agriculture and aquaculture
    Height (English) 16 in.
    Shank Type V-bar molded
    Sole Features Self-cleaning lug outsole for traction in wet, muddy conditions
    Toe Style Plain
    Color Black
    Size 13

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit