Learn More
Dunlop™ Onguard™ Chemical Overboots
Rubber boots for use in emergency response, hazmat environments and site remediation
Manufacturer:  Dunlop™ 870252X00
 View more versions of this product
Catalog No. 19-120-503E
Add to cart
Description
Description
  • 10 in. over-the-boot protection fits over most shoes and boots
  • 4-way cleated sole design for a self-cleaning, slip resistant sole
  • One-piece injection molded construction, eliminates seams for optimal protection
  • Special fold line with non-corrosive snap for snug fit
  • Kick off lug
Specifications
Specifications
Green
Rubber
2X-Large