Learn More
Dunlop™ Onguard™ Chemtex Bib Overalls
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
3X-Large
4X-Large
Description
- constructed of 0.40mm nylon/polyester lining coated with heavy-duty PVC
- Strong, elastic suspenders with durable clips and reinforced stays
- Plain front
- Green
Compliance
Tested to ASTM™ D6413 standard for flame resistance
Specifications
Color | Green |
Material | Nylon, PVC |
Size | Medium |