Learn More
Onguard™ Chemtex Coverall with Attached Hood
Provides Level C protective protection for a variety of chemical applications
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 71020XL00
 View more versions of this product
Catalog No. 19-024535
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM D6413
Attached hood with drawstring and cord lock, reinforced knees, elbows and crotch area, ankle and cuff take up snap, heavy duty zipper and clear ID pocket; PVC on Nylon Polyester
Hooded
X-Large
Protection from a variety of chemical applications
35.5 in.
90.17cm
16.53mil
Zipper
PVC, Nylon, Polyester
Green
Drawstring
78.74cm
31 in.
137.16cm
58 in.