Learn More
Onguard™ Chemtex Jacket with Hood Snaps
Provides Level C chemical protection
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 71032XL00
 View more versions of this product
Catalog No. 19-087-032C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zipper front
Green
Level A environments, chemical, rain, flame
Adjustable waist snap, reinforced knees, elbow and crotch area, snap ankle and cuffs, storm flap front, drawstring with cord lock
0.42mm
Flame
1.0
147.32cm
ASTM D6413
0.4mm Nylon, Polyester Lining Coated with Heavy Duty PVC
X-Large
PVC
Snap
Nylon, Polyester
58 in.