Learn More
Dunlop™ Onguard™ Chest Waders with Cleated Outsole
Superior waterproof protection that resists oil, gas, grease, and saltwater in every condition.
Supplier: Dunlop™ 860671100
Description
These durable chest waders are made of heavy duty PVC and boast cleated outsoles for excellent flexibility and traction clogging prevention, even in wet or muddy terrain.
- Chest high, with PVC upper and non-absorbent polyester backing
- Injection molded, seamless boot construction
- Steel midsole and adjustable polyester and nylon strap
- Excellent cold weather flexibility
- Can be folded or rolled without cracking
- Top drawstring closure for snug fit
- Non-corrosive, reinforced suspender buttons
- Built-in belt loops with adjustable belt
- Replaceable cushion insoles
- Made in the USA
Specifications
ASTM F2413-11 M I/75 C/75 SD | |
Black | |
PVC, Polyester | |
11 | |
Polyester/Nylon |
Drawstring | |
Utilities, Municipalities, Aquaculture, and Commercial Fishing | |
Seamless | |
Cleated Outsole, Steel Midsole, Replaceable Cushion Insole | |
Steel |