Onguard Chest Waders with Cleated Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 860661200

 View more versions of this product

Catalog No. 19-073873

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    Closure Type Top drawstring
    For Use With (Application) For utilities, municipalities, aquaculture and commercial fishing
    Gender Male
    Sole Features Cleated outsole, Steel midsole, replaceable cushion insole
    Strap Material Polyester Nylon
    Toe Style Plain
    Color Black
    Material PVC, Polyester
    Size 12
    Backing Material Polyester
    Includes Reinforced suspender buttons, built-in belt loops
    Seam Type Seamless

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit