Learn More
Dunlop™ Onguard™ Goliath™ PVC Knee Boots
Ideal for general industrial and safety applications
Manufacturer:  Dunlop™ 89680 7
 View more versions of this product
Catalog No. 19-099-168C
Add to cart
Description
Description
  • Constructed of a special urethane/PVC compound with Elvaloy™ and polymeric plasticizers for enhanced strength and high abrasion resistance
  • Superior protection from animal fats (especially poultry), acids, oils, greases, and other chemicals
  • Net-lined for easy on and off, with a removable insole
  • Built-in steel shank for firm arch support
  • Black with a gray Power-Lug outsole
Order Info
Available with or without steel toe in men's sizes only
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications.
Specifications
Specifications
Animal fats, acids, oils, greases and chemicals
Black
Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining
PVC, Urethane
7
Plain
ASTM F2413-05
Pull-on
16 in.
Steel
Gray power-lug outsole and removable insole