Learn More
Dunlop™ Onguard™ Goliath™ PVC Knee Boots
Ideal for general industrial and safety applications
Supplier: Dunlop™ 89682 14
This item is not returnable.
View return policy
Description
- Constructed of a special urethane/PVC compound with Elvaloy™ and polymeric plasticizers for enhanced strength and high abrasion resistance
- Superior protection from animal fats (especially poultry), acids, oils, greases, and other chemicals
- Net-lined for easy on and off, with a removable insole
- Built-in steel shank for firm arch support
- Black with a gray Power-Lug outsole
Order Info
Available with or without steel toe in men's sizes only
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications.
Specifications
ASTM F2413-05 | |
Pull-on | |
16 in. | |
Steel | |
Gray power-lug outsole and removable insole | |
Safety Steel Toe | |
Yes |
Black | |
Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining | |
Urethane, PVC | |
14 | |
Steel | |
Animal fats, acids, oils, greases and chemicals |