Onguard™ Hazmax™ Boots

Choose from two styles

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 870120900

 View more versions of this product

Catalog No. 17-985-403E

  Check Availability
Add to Cart

Alert

Do not use for fire protection.


    Specifications

    Material Flexible PVC alloy
    Size 9
    For Use With (Application) Ideal for emergency response teams, hazmat technicians, environmental specialists, and site remediation workers.
    Color Yellow Outsoles
    Height (English) 16 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit