Onguard Hazmax EZ-Fit 007 Steel Toe Boots

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 87007LG00

 View more versions of this product

Catalog No. 19-161-293

  Check Availability
Add to Cart

    Specifications

    Closure Type Pull-on
    For Use With (Application) Used in situations involving dangerous chemicals, gasses or biological hazards and waste
    Height (English) 11 in.
    Lug Type Kick off
    Shank Material Steel
    Toe Style EZ-fit
    Material PVC
    Size Large
    Certifications/Compliance ASTM F2413-05, CSA Z195
    Gender Male
    Sole Features Non-slip EZ decon outsole, Steel midsole
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit