Learn More
Onguard Hazmax EZ-Fit 007 Steel Toe Boots
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 87007XL00
 View more versions of this product
Catalog No. 19-161-294
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PVC
ASTM F2413-05, CSA Z195
Used in situations involving dangerous chemicals, gasses or biological hazards and waste
11 in.
Steel
EZ-fit
X-Large
Pull-on
Male
Kick off
Non-slip EZ decon outsole, Steel midsole
Black