Learn More
Dunlop™ Onguard™ Hazmax™ EZ-Fit™ Boots
Designed for easy donning and doffing while wearing chemical protective suit
Supplier: Dunlop™ 87015S
Alert:
Caution! Do not use for fire or electrical hazard protection.
Description
- One-piece injection molded with no buttons or elastic bands to break for easier decontamination
- Large kick-off lug for easy removal
- Wider across the instep for easy entry
- Nonslip EZ decon outsole for easier decontamination and cleanup
- Steel toe, steel shank, and steel midsole for complete protection
- Height: 11 in. (28cm)
Order Info
Order normal shoe size–designed to accommodate a fully encapsulated suit. Men's full sizes: Small: 6-8, Medium: 9-10, Large: 11-12, X-Large: 13-15.
Certifications
Certified by S.E.I. to meet the requirements of the NFPA 1991, 2005 Edition Standard on Vapor-Protective ensembles for Hazardous Materials Emergencies.
Compliance
Meets ASTM™ F2413-11; CSA Z195 requirements. NFPA 1991 Standard on Vapor-Protective Ensembles for Hazardous Materials Emergencies and CBRN Terrorism Incidents, 2016.
Specifications
For situation involving dangerous chemicals, gases and waste | |
11 in. | |
28 cm | |
Small |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.