Learn More
Dunlop™ Onguard™ DuraPro™ Chemical Resistant Plain Toe Boots with Steel Shank
High performance compund provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole
Manufacturer:  Dunlop™ 890857
 View more versions of this product
Catalog No. 18-500-504
Add to cart
Description
Description
  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360° pull tab for easy donning
  • Steel shank protects and supports arch
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
  • Composition: Polyblend Upper; Sureflex Outsole
  • Sole Design: Next Generation Safety-Loc
  • Available in sizes ranging from 3 to 16
Specifications
Specifications
Blue
Polyblend, Sureflex
Steel
Next Generation Safety-loc, slip resistant
Chemical resistant Footwear
Male
Chemicals
7
Plain