Learn More
Dunlop™ Onguard™ DuraPro™ XCP Chemical Resistant Steel Toe Boots with Steel Shank
High performance compund provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole
Supplier: Dunlop™ 840868
This item is not returnable.
View return policy
Description
- Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
- 360degree pull tab for easy donning
- Steel shank protects and supports arch
- Larger top circumference for quick and easy pull-on
- 3/4 in. height inner liner allows increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
- Honeycomb design molded inside the heel to reduce impact when walking
- Ergonomically designed foot-bed to provide better underfoot support
- Protective bumpers at rear of boot to prevent premature wear/puncture
- Extended circumference kick-off ledge
- Composition: Superpoly Upper; Sureflex Outsole
- Sole Design: Next Generation Chevron
- Available in sizes ranging from 3 to 16
Specifications
F2413-11 Compliant | |
Superpoly, Sureflex | |
8 | |
Steel | |
Chemical Resistant Footwear |
Brown | |
Steel | |
Next Generation Chevron, slip-resistant | |
Chemicals |