Onguard Men's Flex 3 Work Boots

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 89908110

 View more versions of this product

Catalog No. 19-088-2087E

  Check Availability
Add to Cart

    Specifications

    Shank Material Steel
    Certifications/Compliance ASTM F2413-05
    Closure Type Pull-on
    For Use With (Application) Used in oil rigs, refineries, food processing, general industrial areas, lawn care, dairies, breweries and utilities
    Height (English) 16 in.
    Lug Type Kick off
    Toe Material Steel
    Waterproof Yes
    Material Polyblend PVC, Nitrile
    Size 11
    Gender Male
    Resistant Fats, Oils and chemicals
    Sole Features Grey/yellow power-lug outsole and removable air cushion insole, slip resistant
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit