Onguard Men's Flex-O-Thane PVC Overshoes with Cleated Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 88010SM00

 View more versions of this product

Catalog No. 19-170-486A

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Construction, farming, utilities and food processing
    Gender Male
    Height (English) 4 in.
    Sole Features Self-cleaning, slip resistance, 4 way cleated outsole
    Style Premium
    Color Yellow
    Material PVC, Flex-O-Thane
    Size Small

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit