Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Men's Plain Toe Storm King/Hip Wader Boots with Cleated Outsole
SDP

$180.99

Specifications

Certifications/Compliance ASTM F2413-11
Color Black
Material PVC, Polyester
For Use With (Application) For utilities, municipalities, aquaculture and commercial fishing
Includes Non-absorbent lining
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-024378
SDP
View Documents
Dunlop™
860550800
8
Pack for $180.99
Add to cart
 
19-024375
SDP
View Documents
Dunlop™
860550600
6
Pair for $180.99
Add to cart
 
19-024379
SDP
View Documents
Dunlop™
860550900
9
Pair for $180.99
Add to cart
 
19-024377
SDP
View Documents
Dunlop™
860550700
7
Pair for $180.99
Add to cart
 
19-806-599
SDP
View Documents
Dunlop™
860550013
13
Pair for $180.99
Add to cart
 
Description

Description

Specifications

Specifications

ASTM F2413-11
PVC, Polyester
Non-absorbent lining
Seamless
Polyester Nylon
Black
For utilities, municipalities, aquaculture and commercial fishing
Oil, grease, gasoline, salt water
Cleated outsole, Steel midsole, replaceable cushion insole
Plain
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit