Learn More
Dunlop™ Onguard™ Men's Plain Toe Superpoly Boots with Cleated Outsole
Supplier: Dunlop™ 853011200
This item is not returnable.
View return policy
Specifications
ASTM F2413-05 | |
Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants | |
Kick-off | |
Steel | |
Tan cleated outsole and replaceable cushion insoles | |
Chemicals |
Black | |
16 in. | |
Urethane, PVC | |
12 | |
Plain |