Learn More
Onguard Men's Plain Toe Superpoly Boots with Cleated Outsole
Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 853010600
 View more versions of this product
Catalog No. 19-024319
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ASTM F2413-05
Male
Kick off
Steel
Plain
Urethane, PVC
Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants
16 in.
Chemical
Tan cleated outsole and replaceable cushion insoles
Black
6