Onguard Men's Plain Toe Superpoly Boots with Cleated Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 853010700

 View more versions of this product

Catalog No. 19-024320

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM F2413-05
    For Use With (Application) Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants
    Gender Male
    Height (English) 16 in.
    Lug Type Kick off
    Resistant Chemical
    Shank Material Steel
    Sole Features Tan cleated outsole and replaceable cushion insoles
    Toe Style Plain
    Material Urethane, PVC
    Size 7
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit