Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Men's Plain Toe Terra-Lites Leather Boots with Commando Outsole

Manufacturer:  Dunlop™ 863910900

 View more versions of this product

Catalog No. 19-806-612

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Safety
31.1 x 12.06 cm (12.25 x 4.75 in.)
Freezer lockers and cold storage units, cold weather industrial environments, ice fishing, winter hunting, and snowmobiling
Taslon
31.1 cm
Leather
Commando outsole, Polyurethane bottom
Yes
8.16 kg
12.06 cm
Black
Brass
10 in.
12.25 in.
Thermashield™ 3-ply
9
Plain
18 lb.
4.75 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit