Onguard Men's Slicker Overboots with Self-Cleaning Cleated Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 860500800

 View more versions of this product

Catalog No. 19-051939

  Check Availability
Add to Cart

    Specifications

    Gender Male
    Sole Features Self-cleaning cleated outsole
    Strap Material Polyester
    Color Black
    Size 8
    Buckle Material Plastic
    Height (English) 17 in.
    Liner Material Polyester
    Material PVC, Flex-O-Thane
    For Use With (Application) Construction, concrete work, utilities and food processing

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit