Onguard Men's Steel Toe Comfort NeoMax Boots with Tunnel Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 820021600

 View more versions of this product

Catalog No. 19-806-590

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Certifications/Compliance ASTM F2413-05
    For Use With (Application) Used in variety of applications
    Gender Male
    Height (English) 15 in.
    Shank Material Steel
    Sole Features Tunnel outsole and abrasion resistant outsole, slip resistant
    Color Grey
    Size 16
    Toe Material Steel
    Material PVC

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit