Learn More
Dunlop™ Onguard™ Men's Steel Toe Comfort Polyblend Boots with Tunnel Outsole
Manufacturer:  Dunlop™ 820061400
 View more versions of this product
Catalog No. 19-806-593
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Safety
Black
Used in a variety of applications
PVC
14
Steel
ASTM F2413-05, ANSI Z41 PT99 M I/75 C/75
Pull-on
15 in.
Steel
Tunnel outsole and abrasion-resistant outsole, Slip-resistant