Onguard Men's Steel Toe Comfort Polyblend Boots with Tunnel Outsole

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 820061600

 View more versions of this product

Catalog No. 19-806-594

  Check Availability
Add to Cart

    Specifications

    Toe Material Steel
    Material PVC
    Closure Type Pull-on
    Gender Male
    Height (English) 15 in.
    Shank Material Steel
    Sole Features Tunnel outsole and abrasion resistant outsole, slip resistant
    Color Black
    Size 16
    Certifications/Compliance ASTM F2413-05, ANSI Z41 PT99 M I/75 C/75
    For Use With (Application) Used in a variety of applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit