Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Onguard Men's Therma Toe Terra-Lites Leather Boots with and Commando Outsole

Manufacturer:  Onguard Industries LLCSupplier Diversity Partner 863921200

 View more versions of this product

Catalog No. 19-024288


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Leather
5 in.
ASTM F2413-11
Brass
Male
Taslon
34.5cm
Commando outsole, dual density Polyurethane bottom
Yes
9kg
12
12.7cm
13.5 x 5 in. (34.5 x 12.7cm)
Freezer lockers and cold storage units, cold weather industrial environments, ice fishing, winter hunting, and snowmobiling
10 in.
13.5 in.
Thermashield™ 3-ply with Fleece collar
Therma
20 lb.
Black
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit